missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-butanol 99.0+%, TCI America™

Catalog No. P087625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087625ML Supplier TCI America Supplier No. P087625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used in chemical research and analysis.

Chemical Identifiers

CAS 2344-70-9
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00044349
InChI Key GDWRKZLROIFUML-UHFFFAOYSA-N
Synonym 4-phenyl-2-butanol, 1-phenyl-3-butanol, 2-hydroxy-4-phenylbutane, methyl phenethyl carbinol, methylphenethylcarbinol, 2-butanol, 4-phenyl, alpha-methylbenzenepropanol, phenylethyl methyl carbinol, methyl-2-phenylethylcarbinol
PubChem CID 61302
IUPAC Name 4-phenylbutan-2-ol
SMILES CC(CCC1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 239°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-2-butanol