missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Phenyl-2-butanol, 98%

Catalog No. AAL0597006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0597006 5 g
AAL0597014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0597006 Supplier Thermo Scientific Chemicals Supplier No. L0597006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2344-70-9 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2344-70-9
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00044349
InChI Key GDWRKZLROIFUML-UHFFFAOYSA-N
Synonym 4-phenyl-2-butanol, 1-phenyl-3-butanol, 2-hydroxy-4-phenylbutane, methyl phenethyl carbinol, methylphenethylcarbinol, 2-butanol, 4-phenyl, alpha-methylbenzenepropanol, phenylethyl methyl carbinol, methyl-2-phenylethylcarbinol
PubChem CID 61302
IUPAC Name 4-phenylbutan-2-ol
SMILES CC(CCC1=CC=CC=C1)O

Specifications

Density 0.98
Boiling Point 239°C
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 1908294
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material (+/-)-4-Phenyl-2-butanol
EINECSNumber 219-055-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only