missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-butanol, 97%

Catalog No. AC183400050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC183400050 Supplier Thermo Scientific Chemicals Supplier No. 183400050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2344-70-9 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2344-70-9
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
InChI Key GDWRKZLROIFUML-UHFFFAOYSA-N
Synonym 4-phenyl-2-butanol, 1-phenyl-3-butanol, 2-hydroxy-4-phenylbutane, methyl phenethyl carbinol, methylphenethylcarbinol, 2-butanol, 4-phenyl, alpha-methylbenzenepropanol, phenylethyl methyl carbinol, methyl-2-phenylethylcarbinol
PubChem CID 61302
IUPAC Name 4-phenylbutan-2-ol
SMILES CC(CCC1=CC=CC=C1)O

Specifications

Density 0.9800g/mL
Boiling Point 132.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5157 to 1.5177
Linear Formula C6H5CH2CH2CH(OH)CH3
Quantity 5 g
Beilstein 06,522
Specific Gravity 0.98
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-butanol
Show More Show Less
EINECSNumber 219-055-9

RUO – Research Use Only