missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenyl-1-butene 98.0+%, TCI America™
$117.43 - $337.40
Chemical Identifiers
| CAS | 768-56-9 |
|---|---|
| Molecular Formula | C10H12 |
| Molecular Weight (g/mol) | 132.206 |
| MDL Number | MFCD00026111 |
| InChI Key | PBGVMIDTGGTBFS-UHFFFAOYSA-N |
| PubChem CID | 13033 |
| IUPAC Name | but-3-enylbenzene |
| SMILES | C=CCCC1=CC=CC=C1 |
Chemical Identifiers
| 768-56-9 | |
| 132.206 | |
| PBGVMIDTGGTBFS-UHFFFAOYSA-N | |
| but-3-enylbenzene |
| C10H12 | |
| MFCD00026111 | |
| 13033 | |
| C=CCCC1=CC=CC=C1 |
Specifications
| 768-56-9 | |
| 181°C | |
| MFCD00026111 | |
| 3295 | |
| C=CCCC1=CC=CC=C1 | |
| 132.206 | |
| 132.21 | |
| Liquid |
| Colorless | |
| C10H12 | |
| 5 mL | |
| PBGVMIDTGGTBFS-UHFFFAOYSA-N | |
| but-3-enylbenzene | |
| 13033 | |
| ≥98.0% (GC) | |
| 4-Phenyl-1-butene |
Safety and Handling
EINECSNumber : (3)-4520
RTECSNumber : EM9000000
TSCA : No