missing translation for 'onlineSavingsMsg'
Learn More

4-Phényl-1-butanol 95,0+%, TCI America™

Numéro de catalogue. P12755ML
Modifier l'affichage
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P12755ML 5 mL
P127525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P12755ML Fournisseur TCI America Code fournisseur P12755ML

Identifiants chimiques

CAS 3360-41-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002971
InChI Key LDZLXQFDGRCELX-UHFFFAOYSA-N
Synonym 4-phenyl-1-butanol, benzenebutanol, 4-phenylbutanol, phenylbutyl alcohol, 4-phenylbutyl alcohol, 1-butanol, 4-phenyl, 4-phenyl butanol-1, 4-phenyl butanol, unii-q5orz1321g, 4-phenyl-n-butanol
PubChem CID 76889
IUPAC Name 4-phénylbutan-1-ol
SMILES C1=CC=C(C=C1)CCCCO

Spécifications

Color Jaune
Boiling Point 140°C
Quantity 5 mL
Formula Weight 150.22
Percent Purity ≥95.0% (GC)
Physical Form Liquide
Chemical Name or Material 4-Phenyl-1-butanol
EINECSNumber (3)-1034
TSCA Yes