missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-butanol, 97%

Catalog No. AAA1343330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1343330 250 g
AAA1343309 10 g
AAA1343318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1343330 Supplier Thermo Scientific Chemicals Supplier No. A1343330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3360-41-6 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3360-41-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002971
InChI Key LDZLXQFDGRCELX-UHFFFAOYSA-N
Synonym 4-phenyl-1-butanol, benzenebutanol, 4-phenylbutanol, phenylbutyl alcohol, 4-phenylbutyl alcohol, 1-butanol, 4-phenyl, 4-phenyl butanol-1, 4-phenyl butanol, unii-q5orz1321g, 4-phenyl-n-butanol
PubChem CID 76889
IUPAC Name 4-phenylbutan-1-ol
SMILES C1=CC=C(C=C1)CCCCO

Specifications

Density 0.975
Boiling Point 138°C to 140°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 250 g
Beilstein 2042122
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-butanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-128-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only