missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,3-dioxane 98.0+%, TCI America™

Catalog No. P016625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P016625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P016625G Supplier TCI America Supplier No. P016625G

Chemical Identifiers

CAS 772-00-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00006568
InChI Key RCJRILMVFLGCJY-UHFFFAOYNA-N
Synonym 1,3-dioxane, 4-phenyl, m-dioxane, 4-phenyl, 4-phenyl-m-dioxane, 1,3-dioxane, 4-phenyl-, 4s, acmc-20mb6r, 4-phenyl-1,3 dioxane, acmc-209p8i
PubChem CID 92840
IUPAC Name 4-phenyl-1,3-dioxane
SMILES C1CC(OCO1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1,3-dioxane