missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1,2,4-triazoline-3,5-dione, TCI America™

Catalog No. p-7134993
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11841G 1 g
P11845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11841G Supplier TCI America Supplier No. P11841G
Only null left

Chemical Identifiers

CAS 4233-33-4
Molecular Formula C8H5N3O2
Molecular Weight (g/mol) 175.15
MDL Number MFCD00003148
InChI Key ISULLEUFOQSBGY-UHFFFAOYSA-N
Synonym PTAD
PubChem CID 77913
IUPAC Name 4-phenyl-4,5-dihydro-3H-1,2,4-triazole-3,5-dione
SMILES O=C1N=NC(=O)N1C1=CC=CC=C1

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 175.15
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenyl-1,2,4-triazoline-3,5-dione
TSCA No
Recommended Storage Freezer