missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxypiperidine, 98%

Catalog No. AAH50984MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50984MD 250 mg
AAH5098403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50984MD Supplier Thermo Scientific Chemicals Supplier No. H50984MD

CAS: 3202-33-3 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3202-33-3
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD04114971
InChI Key KBYPITRKIJKGMD-UHFFFAOYSA-N
Synonym piperidine, 4-phenoxy, unii-aof4m4l61z, 4-phenoxy-piperidine, aof4m4l61z, piperidine, 4-phenoxy-, hydrochloride, 4-phenoxy piperidine, acmc-209hq3
PubChem CID 18878
IUPAC Name 4-phenoxypiperidine
SMILES C1CC(CCN1)OC1=CC=CC=C1

Specifications

Boiling Point 107°C to 108°C (0.1 mmHg)
Quantity 250 mg
Formula Weight 177.25
Percent Purity 99%
Chemical Name or Material 4-Phenoxypiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only