missing translation for 'onlineSavingsMsg'
Learn More

(4-Phenoxyphenyl)methylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. CC53763DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53763DA 1g Amber Glass Bottle
CC53763DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53763DA Supplier Thermo Scientific Chemicals Supplier No. CC53763DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169944-04-1
Molecular Formula C13H14ClNO
Molecular Weight (g/mol) 235.711
InChI Key VHCSCKHIGGFTHN-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1
PubChem CID 22293026
IUPAC Name (4-phenoxyphenyl)methanamine;hydrochloride
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 235.71
Percent Purity 97%
Chemical Name or Material (4-Phenoxyphenyl)methylamine hydrochloride
Hazard Category GHS Signal Word: Warning