missing translation for 'onlineSavingsMsg'
Learn More

[4-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC63709CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC63709CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

262862-97-5
214.264
MTQHORUPINPVTP-UHFFFAOYSA-N
11333367
C1=CC=C(C=C1)OCC2=CC=C(C=C2)CO
C14H14O2
MFCD09879941
4-phenoxymethyl phenyl methanol, 4-phenoxymethylphenylmethanol, 4-phenoxymethylbenzyl alcohol, 4-phenoxymethylphenyl-methanol, 4-phenoxymethyl phenyl methan-1-ol
[4-(phenoxymethyl)phenyl]methanol

Specifications

[4-(Phenoxymethyl)phenyl]methanol
96.5
97%
C14H14O2
MFCD09879941
MTQHORUPINPVTP-UHFFFAOYSA-N
[4-(phenoxymethyl)phenyl]methanol
11333367
97%
262862-97-5
100.0
Amber Glass Bottle
250mg
4-phenoxymethyl phenyl methanol, 4-phenoxymethylphenylmethanol, 4-phenoxymethylbenzyl alcohol, 4-phenoxymethylphenyl-methanol, 4-phenoxymethyl phenyl methan-1-ol
C1=CC=C(C=C1)OCC2=CC=C(C=C2)CO
214.264
214.26
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents