missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC63701DA

Quantity 1g

 View more versions of this product

Catalog No. CC63701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

31719-76-3
227.24
GRBUVHSYBRTCIB-UHFFFAOYSA-M
737267
[O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1
C14H11O3
MFCD00623268
4-phenoxymethyl benzoic acid, 4-phenoxymethyl benzenecarboxylic acid, 4-phenoxymethyl-benzoic acid, phenoxymethylbenzenecarboxylicacid
4-(phenoxymethyl)benzoic acid

Specifications

4-(Phenoxymethyl)benzoic acid
96.5
97%
1g
4-phenoxymethyl benzoic acid, 4-phenoxymethyl benzenecarboxylic acid, 4-phenoxymethyl-benzoic acid, phenoxymethylbenzenecarboxylicacid
[O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1
227.24
228.25
31719-76-3
100.0
C14H11O3
MFCD00623268
GRBUVHSYBRTCIB-UHFFFAOYSA-M
4-(phenoxymethyl)benzoic acid
737267
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions