missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenoxymethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 282.75 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(phenoxymethyl)benzenesulfonyl chloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |