missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenoxymethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC63703DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 1002727-89-0 | |
| 282.74 | |
| 4-phenoxymethyl benzenesulfonyl chloride, 4-phenoxymethyl benzene-1-sulfonyl chloride |
| C13H11ClO3S | |
| ITLWVYCIZYWFNW-UHFFFAOYSA-N | |
| 43811043 |
Specifications
| 4-(phenoxymethyl)benzenesulfonyl chloride | |
| Amber Glass Bottle | |
| 1g | |
| ITLWVYCIZYWFNW-UHFFFAOYSA-N | |
| 43811043 | |
| 97% |
| 1002727-89-0 | |
| C13H11ClO3S | |
| 4-phenoxymethyl benzenesulfonyl chloride, 4-phenoxymethyl benzene-1-sulfonyl chloride | |
| 282.74 | |
| 282.75 |
Safety and Handling
GHS Signal Word: Danger