missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC63703CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC63703CB 250mg Amber Glass Bottle
CC63703DA 1g Amber Glass Bottle
CC63703DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC63703CB Supplier Thermo Scientific Chemicals Supplier No. CC63703CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002727-89-0
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.74
InChI Key ITLWVYCIZYWFNW-UHFFFAOYSA-N
Synonym 4-phenoxymethyl benzenesulfonyl chloride, 4-phenoxymethyl benzene-1-sulfonyl chloride
PubChem CID 43811043

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 282.75
Percent Purity 97%
Chemical Name or Material 4-(phenoxymethyl)benzenesulfonyl chloride
Hazard Category GHS Signal Word: Danger