missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC63704CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC63704CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC63704CB Supplier Thermo Scientific Chemicals Supplier No. CC63704CB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2683-70-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD09879939
InChI Key JLGXYDMVIJFOKF-UHFFFAOYSA-N
Synonym 4-phenoxymethyl benzaldehyde, benzaldehyde,4-phenoxymethyl, 4-phenoxymethyl-benzaldehyde
PubChem CID 12106287
IUPAC Name 4-(phenoxymethyl)benzaldehyde
SMILES O=CC1=CC=C(COC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 4-(Phenoxymethyl)benzaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.