missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-2-ol, 97%

Catalog No. AAL0275206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0275206 5 g
AAL0275214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0275206 Supplier Thermo Scientific Chemicals Supplier No. L0275206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2117-11-5 | C7H14OSi | 142.27 g/mol

4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentyn-2-ol is used in Quantitative structure activity relationship (QSAR) studies pertaining to toxic levels of organic compounds in environmental situations. It is also used in the preparation of Fischer-type rhenium(III) benzoyldiazenido-2-oxacyclocarbenes. Further, it is used in the preparation of C-aryl glycosides by undergoing cycloaddition reaction with an aryl nitrile oxide.

Solubility
Miscible with chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 2117-11-5
MDL Number MFCD00004555

Specifications

Density 0.892
Boiling Point 126°C to 127°C
Flash Point 37°C (98°F)
Molecular Formula C7H14OSi
Refractive Index 1.438
Quantity 5 g
UN Number UN1987
Beilstein 1734245
Synonym 4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
Solubility Information Miscible with chloroform and ethyl acetate.
InChI Key VSTMVAFCUIGHQI-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CCCO
IUPAC Name 4-trimethylsilylbut-3-yn-1-ol
Molecular Weight (g/mol) 142.27
PubChem CID 2760827
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material 4-Pentyn-2-ol
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 218-321-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only