missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol 96.0+%, TCI America™

Catalog No. P08175ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08175ML 5 mL
P081725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08175ML Supplier TCI America Supplier No. P08175ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5390-04-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002974
InChI Key CRWVOXFUXPYTRK-UHFFFAOYSA-N
Synonym 4-pentyn-1-ol, 4-pentyne-1-ol, 1-pentyn-5-ol, 5-hydroxypentyne, 4-pentynol, 4-pentin-1-ol, pent-4yn-1-ol, 5-hydroxy-1-pentyne, pent-1-yn-5-ol, pubchem2025
PubChem CID 79346
IUPAC Name pent-4-yn-1-ol
SMILES C#CCCCO

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 84.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pentyn-1-ol