missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyn-1-ol, 95%

Catalog No. AC330430250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330430250 25 g Glass bottle
AC330430050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330430250 Supplier Thermo Scientific Chemicals Supplier No. 330430250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5390-04-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5390-04-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002974
InChI Key CRWVOXFUXPYTRK-UHFFFAOYSA-N
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
PubChem CID 79351
SMILES OCCCC#C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.9100g/mL
Boiling Point 154°C to 155°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC∽C(CH2)3OH
Refractive Index 1.4430 to 1.447
Quantity 25 g
Specific Gravity 0.91
IUPAC Name pent-4-yn-1-ol
Formula Weight 84.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Pentyn-1-ol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-383-6

RUO – Research Use Only.