missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentylbiphenyl 96.0+%, TCI America™
$63.38 - $123.03
Chemical Identifiers
| CAS | 7116-96-3 |
|---|---|
| Molecular Formula | C17H20 |
| Molecular Weight (g/mol) | 224.35 |
| MDL Number | MFCD00009503 |
| InChI Key | IFUOTAQBVGAZPR-UHFFFAOYSA-N |
| Synonym | 4-pentylbiphenyl, 4-n-pentylbiphenyl, 4-pentyl-1,1'-biphenyl, p-pentylbiphenyl, 1,1'-biphenyl, 4-pentyl, 4pentylbiphenyl, 1,1'-biphenyl, pentyl, 4-n-amyldiphenyl, 4-n-amylbiphenyl, pubchem12931 |
| PubChem CID | 81546 |
| IUPAC Name | 4-pentyl-1,1'-biphenyl |
| SMILES | CCCCCC1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 7116-96-3 | |
| 224.35 | |
| IFUOTAQBVGAZPR-UHFFFAOYSA-N | |
| 81546 | |
| CCCCCC1=CC=C(C=C1)C1=CC=CC=C1 |
| C17H20 | |
| MFCD00009503 | |
| 4-pentylbiphenyl, 4-n-pentylbiphenyl, 4-pentyl-1,1'-biphenyl, p-pentylbiphenyl, 1,1'-biphenyl, 4-pentyl, 4pentylbiphenyl, 1,1'-biphenyl, pentyl, 4-n-amyldiphenyl, 4-n-amylbiphenyl, pubchem12931 | |
| 4-pentyl-1,1'-biphenyl |
Specifications
| 7116-96-3 | |
| 167°C | |
| MFCD00009503 | |
| 4-pentylbiphenyl, 4-n-pentylbiphenyl, 4-pentyl-1,1'-biphenyl, p-pentylbiphenyl, 1,1'-biphenyl, 4-pentyl, 4pentylbiphenyl, 1,1'-biphenyl, pentyl, 4-n-amyldiphenyl, 4-n-amylbiphenyl, pubchem12931 | |
| CCCCCC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 224.35 | |
| 224.35 | |
| Liquid |
| Yellow | |
| C17H20 | |
| 5 mL | |
| IFUOTAQBVGAZPR-UHFFFAOYSA-N | |
| 4-pentyl-1,1'-biphenyl | |
| 81546 | |
| ≥96.0% (GC) | |
| 4-Pentylbiphenyl |
Safety and Handling
TSCA : Yes