missing translation for 'onlineSavingsMsg'
Learn More

4'-Pentylacetophenone 95.0+%, TCI America™

Catalog No. P103525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P103525G Supplier TCI America Supplier No. P103525G
Only null left

Chemical Identifiers

CAS 37593-02-5
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00043687
InChI Key KBKGPMDADJLBEM-UHFFFAOYSA-N
Synonym 4'-pentylacetophenone, p-pentylacetophenone, p-n-pentylacetophenone, 1-4-pentylphenyl ethanone, ethanone, 1-4-pentylphenyl, 4'-n-amylacetophenone, 4'-n-pentylacetophenone, acetophenone, 4'-pentyl, 1-4-pentylphenyl ethan-1-one, 1-acetyl-4-pentylbenzene
PubChem CID 98878
IUPAC Name 1-(4-pentylphenyl)ethan-1-one
SMILES CCCCCC1=CC=C(C=C1)C(C)=O

Specifications

Color Colorless
Boiling Point 145°C
Quantity 25 g
Formula Weight 190.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Pentylacetophenone
EINECSNumber (3)-3540
TSCA Yes