missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenenitrile 97.0+%, TCI America™
$385.86 - $1273.14
Chemical Identifiers
| CAS | 592-51-8 |
|---|---|
| Molecular Formula | C5H7N |
| Molecular Weight (g/mol) | 81.12 |
| MDL Number | MFCD00151259 |
| InChI Key | CFEYBLWMNFZOPB-UHFFFAOYSA-N |
| Synonym | 4-pentenenitrile, allylacetonitrile, 4-pentenonitrile, 3-butenyl cyanide, 1-cyano-3-butene, 4-cyano-1-butene, allylmethyl cyanide, 4-pentenoic acid, nitrile, 4-cyano-butene |
| PubChem CID | 11604 |
| IUPAC Name | pent-4-enenitrile |
| SMILES | C=CCCC#N |
Chemical Identifiers
| 592-51-8 | |
| 81.12 | |
| CFEYBLWMNFZOPB-UHFFFAOYSA-N | |
| 11604 | |
| C=CCCC#N |
| C5H7N | |
| MFCD00151259 | |
| 4-pentenenitrile, allylacetonitrile, 4-pentenonitrile, 3-butenyl cyanide, 1-cyano-3-butene, 4-cyano-1-butene, allylmethyl cyanide, 4-pentenoic acid, nitrile, 4-cyano-butene | |
| pent-4-enenitrile |
Specifications
| 592-51-8 | |
| 140°C | |
| MFCD00151259 | |
| 3273 | |
| CFEYBLWMNFZOPB-UHFFFAOYSA-N | |
| pent-4-enenitrile | |
| 11604 | |
| ≥97.0% (GC) | |
| 4-Pentenenitrile |
| Yellow | |
| C5H7N | |
| 5 mL | |
| 4-pentenenitrile, allylacetonitrile, 4-pentenonitrile, 3-butenyl cyanide, 1-cyano-3-butene, 4-cyano-1-butene, allylmethyl cyanide, 4-pentenoic acid, nitrile, 4-cyano-butene | |
| C=CCCC#N | |
| 81.12 | |
| 81.12 | |
| Liquid |
Safety and Handling
TSCA : Yes