missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Penten-2-ol 99.0+%, TCI America™
Supplier: TCI America P180425G
Specifications
| 4-Penten-2-ol | |
| Colorless | |
| C5H10O | |
| 25 g | |
| 4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa | |
| CC(CC=C)O | |
| 86.134 | |
| 86.13 | |
| Liquid |
| 625-31-0 | |
| 116°C | |
| MFCD00004556 | |
| 1987 | |
| ZHZCYWWNFQUZOR-UHFFFAOYSA-N | |
| pent-4-en-2-ol | |
| 12247 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 625-31-0 | |
| 86.134 | |
| ZHZCYWWNFQUZOR-UHFFFAOYSA-N | |
| 12247 | |
| CC(CC=C)O |
| C5H10O | |
| MFCD00004556 | |
| 4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa | |
| pent-4-en-2-ol |
Safety and Handling
EINECSNumber : (2)-2393
TSCA : Yes