missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-2-ol 99.0+%, TCI America™

Catalog No. P18045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18045G 5 g
P180425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18045G Supplier TCI America Supplier No. P18045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-31-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00004556
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYSA-N
Synonym 4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa
PubChem CID 12247
IUPAC Name pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

Color Colorless
Boiling Point 116°C
Quantity 5 g
UN Number 1987
Formula Weight 86.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Penten-2-ol
EINECSNumber (2)-2393
TSCA Yes