missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Penten-2-ol, 98%

CAS: 625-31-0 | C5H10O | 86.134 g/mol

Supplier:  Thermo Scientific Chemicals B2020806

 View more versions of this product

Catalog No. AAB2020806


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-4-Penten-2-ol
0.837
30°C (86°F)
1.424
5 g
4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa
CC(CC=C)O
86.134
86.13
625-31-0
115°C to 116°C
C5H10O
MFCD00004556
UN1987
ZHZCYWWNFQUZOR-UHFFFAOYSA-N
pent-4-en-2-ol
12247
98%

Chemical Identifiers

625-31-0
86.134
ZHZCYWWNFQUZOR-UHFFFAOYSA-N
12247
CC(CC=C)O
C5H10O
MFCD00004556
4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa
pent-4-en-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 210-887-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only