missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Penten-2-ol, 98%

Catalog No. AAB2020803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020803 1 g
AAB2020806 5 g
AAB2020814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2020803 Supplier Thermo Scientific Chemicals Supplier No. B2020803

CAS: 625-31-0 | C5H10O | 86.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-31-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00004556
InChI Key ZHZCYWWNFQUZOR-UHFFFAOYSA-N
Synonym 4-penten-2-ol, 1-penten-4-ol, 4-hydroxypent-1-ene, penten-4-ol, acmc-20apgd, acmc-1bael, 4-emtem-2-ol, ch2=chch2ch oh ch3, zhzcywwnfquzor-uhfffaoysa
PubChem CID 12247
IUPAC Name pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.424
Quantity 1 g
UN Number UN1987
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material (+/-)-4-Penten-2-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 210-887-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only