missing translation for 'onlineSavingsMsg'
Learn More

4-Penten-1-ol, 99%

Catalog No. p-179690
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150010100 10 g Glass bottle
AC150010500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC150010100 Supplier Thermo Scientific Chemicals Supplier No. 150010100
Only null left

CAS: 821-09-0 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 821-09-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002975
InChI Key LQAVWYMTUMSFBE-UHFFFAOYSA-N
Synonym 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
PubChem CID 13181
IUPAC Name pent-4-en-1-ol
SMILES C=CCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8340g/mL
Boiling Point 134°C to 137°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4289 to 1.4309
Linear Formula H2C=CH(CH2)3OH
Quantity 10 g
Beilstein 01, 443
Specific Gravity 0.834
Solubility Information Solubility in water: 57g/L (25°C)
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Penten-1-ol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 212-473-2

RUO – Research Use Only