missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-TEMPO, free radical, 96%

CAS: 2896-70-0 | C9H16NO2

$97.26 - $324.82

Chemical Identifiers

CAS 2896-70-0
Molecular Formula C9H16NO2
MDL Number MFCD00006187
Synonym 4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0108403
View Documents
Thermo Scientific Chemicals
L0108403
1 g
Each for $97.26
Only null left
 
AAL0108406
View Documents
Thermo Scientific Chemicals
L0108406
5 g
Each for $324.82
Only null left
 
Description

Description

4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

Solubility
Soluble in water (partly), acetone, DMSO, 100% ethanol, and diethylether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

2896-70-0
C9H16NO2
MFCD00006187
4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical

Specifications

2896-70-0
C9H16NO2
1 g
4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
170.23
4-Oxo-TEMPO, free radical
36°C to 40°C
MFCD00006187
1818579
Soluble in water (partly),acetone,DMSO,100% ethanol,and diethylether. Insoluble in water.
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 220-778-7

RTECSNumber : TN9200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only