missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-TEMPO, free radical, 96%

Catalog No. AAL0108403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0108403 1 g
AAL0108406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0108403 Supplier Thermo Scientific Chemicals Supplier No. L0108403
Only null left

CAS: 2896-70-0 | C9H16NO2

4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Oxo-Tempo, Free radical is used as a useful Spin Trap for in Vitro Experiments. Reaction Between Tempone And Biologically Significant Reactive Oxygen Species can be monitored By Esr Spectroscopy. It is also used In the synthesis And Reactions Of Stable Nitroxyl Radicals.

Solubility
Soluble in water (partly), acetone, DMSO, 100% ethanol, and diethylether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 2896-70-0
Molecular Formula C9H16NO2
MDL Number MFCD00006187
Synonym 4-Oxo-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical

Specifications

Melting Point 36°C to 40°C
Quantity 1 g
Beilstein 1818579
Solubility Information Soluble in water (partly),acetone,DMSO,100% ethanol,and diethylether. Insoluble in water.
Formula Weight 170.23
Percent Purity 96%
Chemical Name or Material 4-Oxo-TEMPO, free radical
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 220-778-7
RTECSNumber TN9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only