missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-L-proline hydrobromide, 90+%

CAS: 75776-67-9 | C5H8BrNO3 | 210.027 g/mol

Supplier:  Thermo Scientific Chemicals H5769103

Quantity 1 g

 View more versions of this product

Catalog No. AAH5769103

Only null left

Description

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

75776-67-9
210.027
SFSROLZHCPCODJ-WCCKRBBISA-N
12773661
C1C(NCC1=O)C(=O)O.Br
C5H8BrNO3
MFCD12024572
4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide
(2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide

Specifications

134°C to 140°C
75776-67-9
MFCD12024572
SFSROLZHCPCODJ-WCCKRBBISA-N
(2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide
210.027
210.03
≥90%
4-Oxo-L-proline hydrobromide
C5H8BrNO3
4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide
C1C(NCC1=O)C(=O)O.Br
1 g
12773661
−32° (c=0.5 in Methanol)
Moisture sensitive
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions