missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4940-39-0
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
MDL Number MFCD00006838
InChI Key RVMGXWBCQGAWBR-UHFFFAOYSA-N
Synonym chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
PubChem CID 2741
IUPAC Name 4-oxochromene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC123880050
SDS View Documents
Thermo Scientific Chemicals
123880050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC123880250
SDS View Documents
Thermo Scientific Chemicals
123880250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4940-39-0
190.15
RVMGXWBCQGAWBR-UHFFFAOYSA-N
2741
C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
C10H6O4
MFCD00006838
chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
4-oxochromene-2-carboxylic acid

Specifications

4940-39-0
100.0
97%
C10H6O4
5g
15, 2241
Solubility in water: sparingly soluble. Other solubilities: soluble in ammonia,soluble in alcohol
C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O
190.15
190.15
4-Oxo-4H-1-benzopyran-2-carboxylic acid, 97%
96.0
Authentic
Glass bottle
MFCD00006838
18, 428
chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
RVMGXWBCQGAWBR-UHFFFAOYSA-N
4-oxochromene-2-carboxylic acid
2741
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-583-0

RTECSNumber : DJ2476000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only