missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-oxo-4-(2-thienyl)butanoic acid, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB10644EB
Specifications
| 4-Oxo-4-(2-thienyl)butanoic acid | |
| 97% | |
| C8H8O3S | |
| 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
| C1=CSC(=C1)C(=O)CCC(=O)O | |
| 184.21 | |
| CHEBI:64434 | |
| 97% |
| 4653-08-1 | |
| Amber glass bottle | |
| 25g | |
| ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
| 4-oxo-4-thiophen-2-ylbutanoic acid | |
| 78385 | |
| 184.21 |
Chemical Identifiers
| 4653-08-1 | |
| 184.21 | |
| 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
| CHEBI:64434 | |
| C1=CSC(=C1)C(=O)CCC(=O)O |
| C8H8O3S | |
| ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
| 78385 | |
| 4-oxo-4-thiophen-2-ylbutanoic acid |
Safety and Handling
GHS Signal Word: Warning