missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 95.0+%, TCI America™
$188.27 - $617.00
Chemical Identifiers
| CAS | 2896-70-0 |
|---|---|
| Molecular Formula | C9H16NO2 |
| MDL Number | MFCD00006187 |
| Synonym | 4-Oxo-TEMPO |
Chemical Identifiers
| 2896-70-0 | |
| C9H16NO2 | |
| MFCD00006187 | |
| 4-Oxo-TEMPO |
Specifications
| 2896-70-0 | |
| C9H16NO2 | |
| 5 g | |
| 4-Oxo-TEMPO | |
| ≥95.0% (GC,T) | |
| 4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
| 37°C | |
| MFCD00006187 | |
| 3077 | |
| 170.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TN9200000
TSCA : Yes
Recommended Storage : Freezer