missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 37°C |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 170.23 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Oxo-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
Safety and Handling
| RTECSNumber | TN9200000 |
| TSCA | Yes |
| Recommended Storage | Freezer |