missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-1-phenylcyclohexanecarbonitrile 98.0+%, TCI America™
$256.05 - $256.05
Chemical Identifiers
| CAS | 25115-74-6 |
|---|---|
| Molecular Formula | C13H13NO |
| Molecular Weight (g/mol) | 199.25 |
| MDL Number | MFCD00044816 |
| InChI Key | GKXOABVSZWCJJK-UHFFFAOYSA-N |
| Synonym | 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone |
| PubChem CID | 91282 |
| IUPAC Name | 4-oxo-1-phenylcyclohexane-1-carbonitrile |
| SMILES | O=C1CCC(CC1)(C#N)C1=CC=CC=C1 |
Chemical Identifiers
| 25115-74-6 | |
| 199.25 | |
| GKXOABVSZWCJJK-UHFFFAOYSA-N | |
| 91282 | |
| O=C1CCC(CC1)(C#N)C1=CC=CC=C1 |
| C13H13NO | |
| MFCD00044816 | |
| 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone | |
| 4-oxo-1-phenylcyclohexane-1-carbonitrile |
Specifications
| 25115-74-6 | |
| White | |
| MFCD00044816 | |
| 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone | |
| O=C1CCC(CC1)(C#N)C1=CC=CC=C1 | |
| 199.25 | |
| 199.25 | |
| Crystalline Powder |
| 119°C | |
| C13H13NO | |
| 5 g | |
| GKXOABVSZWCJJK-UHFFFAOYSA-N | |
| 4-oxo-1-phenylcyclohexane-1-carbonitrile | |
| 91282 | |
| ≥98.0% (GC) | |
| 4-Oxo-1-phenylcyclohexanecarbonitrile |
Safety and Handling
TSCA : No