missing translation for 'onlineSavingsMsg'
Learn More

4-Oxazolecarboxylic Acid 98.0+%, TCI America™

Catalog No. O02885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02885G 5 g
O02881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02885G Supplier TCI America Supplier No. O02885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23012-13-7
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD06797151
InChI Key JBCFJMYPJJWIRG-UHFFFAOYSA-N
Synonym oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 4-oxazolecarboxylicacid, 4-carboxy-1,3-oxazole, oxazole-4-carboxylic, 4-oxazolecarboxlic acid, pubchem11025, 4-carboxyoxazole, 4-oxazolecarboxylic acid ep
PubChem CID 14281430
IUPAC Name 1,3-oxazole-4-carboxylic acid
SMILES C1=C(N=CO1)C(=O)O

Specifications

Melting Point 142°C
Quantity 5 g
Formula Weight 113.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Oxazolecarboxylic Acid