missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 98+%

CAS: 1942-45-6 | C8H14 | 110.2 g/mol

$92.36 - $92.36

Chemical Identifiers

CAS 1942-45-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00009471
InChI Key GZTNBKQTTZSQNS-UHFFFAOYSA-N
Synonym 4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
PubChem CID 16029
IUPAC Name oct-4-yne
SMILES CCCC#CCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223520100
SDS View Documents
Thermo Scientific Chemicals
223520100
10 mL Glass bottle
Each for $92.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1942-45-6
110.2
GZTNBKQTTZSQNS-UHFFFAOYSA-N
16029
CCCC#CCCC
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
oct-4-yne

Specifications

1942-45-6
100.0
Colorless to Yellow
131.0°C to 132.0°C
Authentic
Glass bottle
1.4240 to 1.4260
MFCD00009471
01, III, 1006
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
CCCC#CCCC
110.2
110.2
Liquid
98.5
-103.0°C
0.7500g/mL
29°C
98% min. (GC)
C8H14
CH3CH2CH2C≡CCH2CH2CH3
10 mL
0.75
GZTNBKQTTZSQNS-UHFFFAOYSA-N
oct-4-yne
16029
98+%
4-Octyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 217-730-2

SDS
Documents

Documents

RUO – Research Use Only