missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 98+%

Catalog No. AC223520100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223520100 10 mL Glass bottle
AC223520500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223520100 Supplier Thermo Scientific Chemicals Supplier No. 223520100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1942-45-6 | C8H14 | 110.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1942-45-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00009471
InChI Key GZTNBKQTTZSQNS-UHFFFAOYSA-N
Synonym 4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
PubChem CID 16029
IUPAC Name oct-4-yne
SMILES CCCC#CCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -103.0°C
Color Colorless to Yellow
Density 0.7500g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260
Linear Formula CH3CH2CH2C≡CCH2CH2CH3
Quantity 10 mL
Beilstein 01, III, 1006
Specific Gravity 0.75
Formula Weight 110.2
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Octyne
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 217-730-2

RUO – Research Use Only