Learn More
4-Octyne, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223520500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 4-Octyne | |
| 98.5 | |
| 0.7500g/mL | |
| 29°C | |
| 98% min. (GC) | |
| C8H14 | |
| CH3CH2CH2C≡CCH2CH2CH3 | |
| 50mL | |
| 0.75 | |
| GZTNBKQTTZSQNS-UHFFFAOYSA-N | |
| oct-4-yne | |
| 16029 | |
| 98+% |
| 1942-45-6 | |
| 100.0 | |
| 131.0°C to 132.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4240 to 1.4260 | |
| MFCD00009471 | |
| 01, III, 1006 | |
| 4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide | |
| CCCC#CCCC | |
| 110.2 | |
| 110.2 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 217-730-2
RUO â Research Use Only