missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrosoresorcinol 1-Monomethyl Ether 98.0+%, TCI America™

Catalog No. p-7133717
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
Catalog No. Quantity
N02711G 1 g
N027125G 25 g
2 options

Catalog No. N02711G

Supplier: TCI America N02711G

Only null left

Chemical Identifiers

CAS 13895-38-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00059952
InChI Key RNBFLAJNUZQUIK-UHFFFAOYSA-N
Synonym Coniferron, 4-Methoxy-1,2-benzoquinone 1-Monoxime, 2-Hydroxy-4-methoxy-1-nitrosobenzene, 5-Methoxy-2-nitrosophenol
PubChem CID 83801
IUPAC Name 5-methoxy-2-nitrosophenol
SMILES COC1=CC(=C(C=C1)N=O)O

Specifications

Melting Point 156°C
Quantity 1 g
UN Number 1325
Formula Weight 153.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrosoresorcinol 1-Monomethyl Ether
RTECSNumber SL7805000
TSCA Yes