missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitroquinoline N-Oxide 98.0+%, TCI America™
$202.15 - $623.75
Chemical Identifiers
| CAS | 56-57-5 |
|---|---|
| Molecular Formula | C9H6N2O3 |
| Molecular Weight (g/mol) | 190.16 |
| MDL Number | MFCD00006738 |
| InChI Key | YHQDZJICGQWFHK-UHFFFAOYSA-N |
| Synonym | 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide |
| PubChem CID | 5955 |
| ChEBI | CHEBI:16907 |
| IUPAC Name | 4-nitroquinolin-1-ium-1-olate |
| SMILES | [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1 |
Chemical Identifiers
| 56-57-5 | |
| 190.16 | |
| YHQDZJICGQWFHK-UHFFFAOYSA-N | |
| 5955 | |
| 4-nitroquinolin-1-ium-1-olate |
| C9H6N2O3 | |
| MFCD00006738 | |
| 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide | |
| CHEBI:16907 | |
| [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1 |
Specifications
| 56-57-5 | |
| C9H6N2O3 | |
| 1 g | |
| YHQDZJICGQWFHK-UHFFFAOYSA-N | |
| 4-nitroquinolin-1-ium-1-olate | |
| 5955 | |
| 190.16 | |
| Crystalline Powder |
| 156°C | |
| MFCD00006738 | |
| 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide | |
| [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1 | |
| 190.16 | |
| CHEBI:16907 | |
| ≥98.0% (HPLC,T) | |
| 4-Nitroquinoline N-Oxide |
Safety and Handling
RTECSNumber : VC2100000
TSCA : No