missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroquinoline N-Oxide 98.0+%, TCI America™

Catalog No. N02505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02505G 5 g
N02501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02505G Supplier TCI America Supplier No. N02505G
Only null left

Chemical Identifiers

CAS 56-57-5
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.16
MDL Number MFCD00006738
InChI Key YHQDZJICGQWFHK-UHFFFAOYSA-N
Synonym 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide
PubChem CID 5955
ChEBI CHEBI:16907
IUPAC Name 4-nitroquinolin-1-ium-1-olate
SMILES [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1

Specifications

Melting Point 156°C
Quantity 5 g
Formula Weight 190.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitroquinoline N-Oxide
RTECSNumber VC2100000
TSCA No