missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyridine N-Oxide 98.0+%, TCI America™

Catalog No. N041025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N041025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N041025G Supplier TCI America Supplier No. N041025G

Chemical Identifiers

CAS 1124-33-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00006206
InChI Key RXKNNAKAVAHBNK-UHFFFAOYSA-N
Synonym 4-nitropyridine n-oxide, 4-nitropyridine 1-oxide, 4-nitropyridine-n-oxide, 4-nitropyridine-1-oxide, pyridine, 4-nitro-, 1-oxide, 4-nitropyridin-1-ium-1-olate, p-nitropyridine n-oxide, 4-nitropyridine oxide, 4-nitro-n-oxopyridine, 4-nitro-1-oxido-pyridin-1-ium
PubChem CID 14300
IUPAC Name 4-nitropyridin-1-ium-1-olate
SMILES [O-][N+](=O)C1=CC=[N+]([O-])C=C1

Specifications

Melting Point 161°C
Quantity 25 g
UN Number 2811
Formula Weight 140.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitropyridine N-Oxide
RTECSNumber UT6380000
TSCA Yes