missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyridine N-oxide, 97%, Thermo Scientific™

Catalog No. p-174606
Change view
Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128920050 5g Glass bottle
AC128921000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128920050 Supplier Thermo Scientific Chemicals Supplier No. 128920050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1124-33-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00006206
InChI Key RXKNNAKAVAHBNK-UHFFFAOYSA-N
Synonym 4-nitropyridine n-oxide, 4-nitropyridine 1-oxide, 4-nitropyridine-n-oxide, 4-nitropyridine-1-oxide, pyridine, 4-nitro-, 1-oxide, 4-nitropyridin-1-ium-1-olate, p-nitropyridine n-oxide, 4-nitropyridine oxide, 4-nitro-n-oxopyridine, 4-nitro-1-oxido-pyridin-1-ium
PubChem CID 14300
IUPAC Name 4-nitro-1-oxidopyridin-1-ium
SMILES [O-][N+](=O)C1=CC=[N+]([O-])C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5g
Solubility Information Solubility in water: insoluble
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material 4-Nitropyridine N-oxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 214-395-4
RTECSNumber UT6380000
TSCA TSCA

RUO – Research Use Only