missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyridine N-oxide, 97%

Catalog No. p-7025745
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AAB2140414 25 g
AAB2140422 100 g
2 options

Catalog No. AAB2140414

Supplier: Thermo Scientific Chemicals B2140414

Only null left

CAS: 1124-33-0 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1124-33-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00006206
InChI Key RXKNNAKAVAHBNK-UHFFFAOYSA-N
Synonym 4-nitropyridine n-oxide, 4-nitropyridine 1-oxide, 4-nitropyridine-n-oxide, 4-nitropyridine-1-oxide, pyridine, 4-nitro-, 1-oxide, 4-nitropyridin-1-ium-1-olate, p-nitropyridine n-oxide, 4-nitropyridine oxide, 4-nitro-n-oxopyridine, 4-nitro-1-oxido-pyridin-1-ium
PubChem CID 14300
SMILES [O-][N+](=O)C1=CC=[N+]([O-])C=C1

Specifications

Melting Point 158°C to 163°C
Quantity 25 g
UN Number UN2811
Beilstein 124331
IUPAC Name 4-nitro-1-oxidopyridin-1-ium
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material 4-Nitropyridine N-oxide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H300-H341-H315-H319-H335
Fatal if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-395-4
RTECSNumber UT6380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only