missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$57.75 - $57.75
Chemical Identifiers
| CAS | 24067-17-2 |
|---|---|
| Molecular Formula | C6H6BNO4 |
| Molecular Weight (g/mol) | 166.93 |
| MDL Number | MFCD00161360 |
| InChI Key | NSFJAFZHYOAMHL-UHFFFAOYSA-N |
| PubChem CID | 2773552 |
| IUPAC Name | (4-nitrophenyl)boronic acid |
| SMILES | OB(O)C1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 24067-17-2 | |
| 166.93 | |
| NSFJAFZHYOAMHL-UHFFFAOYSA-N | |
| (4-nitrophenyl)boronic acid |
| C6H6BNO4 | |
| MFCD00161360 | |
| 2773552 | |
| OB(O)C1=CC=C(C=C1)[N+]([O-])=O |
Specifications
| 24067-17-2 | |
| White-Yellow | |
| MFCD00161360 | |
| 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene | |
| OB(O)C1=CC=C(C=C1)[N+]([O-])=O | |
| 166.93 | |
| 166.93 | |
| 4-Nitrophenylboronic Acid (contains varying amounts of Anhydride) |
| 305°C | |
| C6H6BNO4 | |
| 1 g | |
| NSFJAFZHYOAMHL-UHFFFAOYSA-N | |
| (4-nitrophenyl)boronic acid | |
| 2773552 | |
| Crystalline Powder |
Safety and Handling
TSCA : No