missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. N08121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08121G Supplier TCI America Supplier No. N08121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24067-17-2
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161360
InChI Key NSFJAFZHYOAMHL-UHFFFAOYSA-N
PubChem CID 2773552
IUPAC Name (4-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 305°C
Color White-Yellow
Quantity 1 g
Synonym 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
Formula Weight 166.93
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylboronic Acid (contains varying amounts of Anhydride)