Learn More
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol
Supplier: Thermo Scientific Chemicals A1231014
Description
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSubstrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
Solubility
Miscible with water. Immiscible with ethanol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
| 4-Nitrophenyl phosphate disodium salt hexahydrate | |
| >300°C | |
| MFCD00007319 | |
| 5324661 | |
| Miscible with water. Immiscible with ethanol and acetone. | |
| [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 | |
| 263.05 | |
| 371.15 (263.06 Anhydrous) |
| 333338-18-4 | |
| C6H4NNa2O6P | |
| 25 g | |
| pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
| VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
| disodium;(4-nitrophenyl) phosphate | |
| 77949 | |
| 98% |
Chemical Identifiers
| 333338-18-4 | |
| 263.05 | |
| VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
| 77949 | |
| [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
| C6H4NNa2O6P | |
| MFCD00007319 | |
| pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
| disodium;(4-nitrophenyl) phosphate |
Safety and Handling
EINECSNumber : 224-246-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only