missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals

Catalog No. AAJ61401KN
Change view
Click to view available options
Quantity:
100 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61401KN 100 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61401KN Supplier Thermo Scientific Chemicals Supplier No. J61401KN
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for alkaline and acid phosphatase determinations

4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

Solubility
It is soluble in water.

Notes
The tablets should be stored -20°C

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Color Yellow
Quantity 100 Pc.
Beilstein 5324661
Solubility Information It is soluble in water.
Formula Weight 371.15 (263.05 Anhydrous)
Physical Form Tablets
Chemical Name or Material 4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets
EINECSNumber 224-246-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only