missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl Phosphate Di(tris) Salt Hydrate 90.0+%, TCI America™
$134.75 - $134.75
Chemical Identifiers
| CAS | 68189-42-4 |
|---|---|
| Molecular Formula | C14H28N3O12P |
| Molecular Weight (g/mol) | 461.361 |
| MDL Number | MFCD00037009 |
| InChI Key | XXAXKCWOTRABOW-UHFFFAOYSA-N |
| Synonym | Phosphoric Acid 4-Nitrophenyl Ester Bis[tris(hydroxymethyl)aminomethane] Salt, 4-Nitrophenyl Phosphate Bis(tris) Salt |
| PubChem CID | 109721 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;(4-nitrophenyl) dihydrogen phosphate |
| SMILES | C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O.C(C(CO)(CO)N)O.C(C(CO)(CO)N)O |
Chemical Identifiers
| 68189-42-4 | |
| 461.361 | |
| XXAXKCWOTRABOW-UHFFFAOYSA-N | |
| 109721 | |
| C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O.C(C(CO)(CO)N)O.C(C(CO)(CO)N)O |
| C14H28N3O12P | |
| MFCD00037009 | |
| Phosphoric Acid 4-Nitrophenyl Ester Bis[tris(hydroxymethyl)aminomethane] Salt, 4-Nitrophenyl Phosphate Bis(tris) Salt | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;(4-nitrophenyl) dihydrogen phosphate |
Specifications
| 68189-42-4 | |
| MFCD00037009 | |
| Phosphoric Acid 4-Nitrophenyl Ester Bis[tris(hydroxymethyl)aminomethane] Salt, 4-Nitrophenyl Phosphate Bis(tris) Salt | |
| C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O.C(C(CO)(CO)N)O.C(C(CO)(CO)N)O | |
| 461.361 | |
| 461.36 | |
| Crystalline Powder |
| C14H28N3O12P | |
| 5 g | |
| XXAXKCWOTRABOW-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;(4-nitrophenyl) dihydrogen phosphate | |
| 109721 | |
| ≥90.0% (T) | |
| 4-Nitrophenyl Phosphate Di(tris) Salt Hydrate [Substrate for Phosphatase] |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer